5-Amino-2,2-dimethyl-pentan-1-ol 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

OR12007-1G
APOSOR12007-1GEA 66.2 GBP
APOSOR12007-1G
5-Amino-2,2-dimethyl-pentan-1-ol 98%
5-Amino-2,2-dimethyl-pentan-1-ol
Formula: C₇H₁₇NO
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00043626
CAS Number: 13532-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR