2-(Morpholinosulphonyl)phenylboronic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR11971-100MG
APOSOR11971-100MGEA 15.8 GBP
APOSOR11971-100MG
2-(Morpholinosulphonyl)phenylboronic acid
2-(Morpholinosulphonyl)phenylboronic acid
Formula: C₁₀H₁₄BNO₅S
Storage Temperature: Ambient
MDL Number: MFCD09878327
CAS Number: 957062-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR