6-Propyl-2-thiouracil 97+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Propylthiouracil, 4-Hydroxy-2-mercapto-6-propylpyrimidine, 4-Propyl-2-thiouracil, 6-Propyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one

OR11311-100G OR11311-25G
APOSOR11311-100GEA 0 GBP
APOSOR11311-100G APOSOR11311-25G
6-Propyl-2-thiouracil 97+%
6-Propyl-2-thiouracil
Formula: C₇H₁₀N₂OS
Melting Pt: 218…220 °C
Flash Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006041
CAS Number: 51-52-5
EINECS: 200-103-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR