Pyrogallol 98+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Benzene-1,2,3-triol, 1,2,3-Trihydroxybenzene

OR11265-100G OR11265-1KG OR11265-500G
APOSOR11265-100GEA 15.8 GBP
APOSOR11265-100G APOSOR11265-1KG APOSOR11265-500G
Pyrogallol 98+%
Pyrogallol
Formula: C₆H₆O₃
Boiling Pt: 309 °C
Melting Pt: 131…135 °C
Density: 1.45 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002192
CAS Number: 87-66-1
EINECS: 201-762-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR