N-(4-Chloro-3-nitrophenyl)-1H-pyrazole-1-carboxamide

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR111082-1G OR111082-5G OR111082-10G
APOSOR111082-1GEA 0 GBP
APOSOR111082-1G APOSOR111082-5G APOSOR111082-10G
N-(4-Chloro-3-nitrophenyl)-1H-pyrazole-1-carboxamide
N-(4-Chloro-3-nitrophenyl)-1H-pyrazole-1-carboxamide
Formula: C₁₀H₇ClN₄O₃
Storage Temperature: Ambient
MDL Number: MFCD28347983
CAS Number: 1858249-82-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR