1-(5-Chloro-1-benzofuran-2-yl)ethanone

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR110910-1G OR110910-250MG
APOSOR110910-1GEA 126 GBP
APOSOR110910-1G APOSOR110910-250MG
1-(5-Chloro-1-benzofuran-2-yl)ethanone
1-(5-Chloro-1-benzofuran-2-yl)ethanone
Formula: C₁₀H₇ClO₂
Storage Temperature: Ambient
MDL Number: MFCD00661909
CAS Number: 1646-32-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR