Tetrahydro-2H-pyran-4-yl methanesulphonate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 4-Tetrahydropyranyl methanesulphonate

OR110580-1G OR110580-5G
APOSOR110580-1GEA 15.8 GBP
APOSOR110580-1G APOSOR110580-5G
Tetrahydro-2H-pyran-4-yl methanesulphonate
Tetrahydro-2H-pyran-4-yl methanesulphonate
Formula: C₆H₁₂O₄S
Melting Pt: 46…48 °C
Storage Temperature: Ambient
MDL Number: MFCD11505063
CAS Number: 134419-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR