Ethyl pyrazine-2-carboxylate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR110404-1G OR110404-5G OR110404-250MG
APOSOR110404-1GEA 27.3 GBP
APOSOR110404-1G APOSOR110404-5G APOSOR110404-250MG
Ethyl pyrazine-2-carboxylate
Ethyl pyrazine-2-carboxylate
Formula: C₇H₈N₂O₂
Boiling Pt: 120 °C/6mmHg
Melting Pt: 49…52 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00792887
CAS Number: 6924-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR