4-(1H-Pyrazol-3-yl)pyridine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR110289-1G
APOSOR110289-1GEA 41 GBP
APOSOR110289-1G
4-(1H-Pyrazol-3-yl)pyridine
4-(1H-Pyrazol-3-yl)pyridine
Formula: C₈H₇N₃
Melting Pt: 157…158 °C
Density: 1.391 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00115150
CAS Number: 17784-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR