1H-Pyrazole-4-boronic acid pinacol ester

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

OR11023-5G OR11023-25G OR11023-100G
APOSOR11023-5GEA 15.8 GBP
APOSOR11023-5G APOSOR11023-25G APOSOR11023-100G
1H-Pyrazole-4-boronic acid pinacol ester
1H-Pyrazole-4-boronic acid pinacol ester
Formula: C₉H₁₅BN₂O₂
Melting Pt: 142…149 °C
Storage Temperature: Ambient
MDL Number: MFCD03453063
CAS Number: 269410-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR