(4-Aminophenyl)acetic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 4-Aminophenylacetic acid

OR10955-500G
APOSOR10955-500GEA 167 GBP
APOSOR10955-500G
(4-Aminophenyl)acetic acid
(4-Aminophenyl)acetic acid
Formula: (C₆H₄(NH₂))CH₂COOH
Melting Pt: 201 °C
Storage Temperature: Ambient
MDL Number: MFCD00007916
CAS Number: 1197-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR