18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 1,4,7,10,13,16-Hexaoxacyclooctadecane, 18-Crown-6

OR10919-100G OR10919-25G
APOSOR10919-100GEA 17.9 GBP
APOSOR10919-100G APOSOR10919-25G
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) 98%
18-Crown-6 (1,4,7,10,13,16-Hexaoxacyclooctadecane)
Formula: C₁₂H₂₄O₆
MW: 264.32 g/mol
Boiling Pt: 116 °C/0.2mmHg
Melting Pt: 37…45 °C
Density: 1.175 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00005113
CAS Number: 17455-13-9
EINECS: 241-473-5
Merck Index: 13,02630

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR