2,5-Dichlorobenzaldehyde 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR10904-5G OR10904-25G OR10904-100G
APOSOR10904-5GEA 15.8 GBP
APOSOR10904-5G APOSOR10904-25G APOSOR10904-100G
2,5-Dichlorobenzaldehyde 98%
2,5-Dichlorobenzaldehyde
Formula: C₇H₄Cl₂O
Boiling Pt: 231…233 °C
Melting Pt: 53…58 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00156140
CAS Number: 6361-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR