2',4'-Dihydroxy-3'-methylpropiophenone

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR10854-1G OR10854-5G
APOSOR10854-1GEA 184 GBP
APOSOR10854-1G APOSOR10854-5G
2',4'-Dihydroxy-3'-methylpropiophenone
2',4'-Dihydroxy-3'-methylpropiophenone
Formula: C₁₀H₁₂O₃
Melting Pt: 124…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00010818
CAS Number: 63876-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR