Dimethyl hydrogen phosphate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

OR10807-1G OR10807-5G
APOSOR10807-1GEA 47.3 GBP
APOSOR10807-1G APOSOR10807-5G
Dimethyl hydrogen phosphate
Dimethyl hydrogen phosphate
Formula: C₂H₇O₄P
Boiling Pt: 174 °C
Density: 1.323 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS Number: 813-78-5
EINECS: 212-389-6
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR