Tetraethyl methylenebisphosphonate 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Tetraethyl methylenediphosphonate, Tetraethyl methylenebis(phosphonate)

OR10799-1G OR10799-25G OR10799-5G
APOSOR10799-1GEA 15.8 GBP
APOSOR10799-1G APOSOR10799-25G APOSOR10799-5G
Tetraethyl methylenebisphosphonate 97%
Tetraethyl methylenebisphosphonate
Formula: C₉H₂₂O₆P₂
MW: 288.22 g/mol
Boiling Pt: 171…174 °C
Density: 1.162 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00039887
CAS Number: 1660-94-2
EINECS: 216-764-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR