4-(4-Morpholinylcarbonyl)phenylboronic acid 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 4-(4-Morpholinylcarbonyl)benzeneboronic acid, (4-(Morpholine-4-carbonyl)phenyl)boronic acid

OR10568-1G OR10568-5G OR10568-25G
APOSOR10568-1GEA 86.5 GBP
APOSOR10568-1G APOSOR10568-5G APOSOR10568-25G
4-(4-Morpholinylcarbonyl)phenylboronic acid 98%
4-(4-Morpholinylcarbonyl)phenylboronic acid
Formula: C₁₁H₁₄BNO₄
Melting Pt: 126…132 °C
Storage Temperature: Ambient
MDL Number: MFCD03411952
CAS Number: 389621-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR