1-Methylimidazole-5-carboxylic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 1-Methylimidazole-5-carboxylic Acid

OR10543-1G OR10543-25G OR10543-5G
APOSOR10543-1GEA 15.8 GBP
APOSOR10543-1G APOSOR10543-25G APOSOR10543-5G
1-Methylimidazole-5-carboxylic acid
1-Methylimidazole-5-carboxylic acid
Formula: C₅H₆N₂O₂
Melting Pt: 247…250 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00955677
CAS Number: 41806-40-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR