4-Bromophenyl methyl sulphone 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR10386-1G
APOSOR10386-1GEA 15.8 GBP
APOSOR10386-1G
4-Bromophenyl methyl sulphone 98%
4-Bromophenyl methyl sulfone
Formula: C₇H₇BrO₂S
Melting Pt: 101…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00025065
CAS Number: 3466-32-8
EINECS: 222-421-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR