4-(Bromomethyl)tetrahydropyran 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

OR10382-1G OR10382-25G OR10382-5G
APOSOR10382-1GEA 15.8 GBP
APOSOR10382-1G APOSOR10382-25G APOSOR10382-5G
4-(Bromomethyl)tetrahydropyran 97%
4-(Bromomethyl)tetrahydropyran
Formula: C₆H₁₁BrO
MW: 179.0549 g/mol
Boiling Pt: 89…93 °C/10mmHg
Storage Temperature: Freezer
MDL Number: MFCD03788561
CAS Number: 125552-89-8
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR