3-(Chloromethyl)-5-(3-methoxyphenyl)-1,2,4-oxadiazole

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

OR103361-1G OR103361-250MG
APOSOR103361-1GEA 24.2 GBP
APOSOR103361-1G APOSOR103361-250MG
3-(Chloromethyl)-5-(3-methoxyphenyl)-1,2,4-oxadiazole
3-(Chloromethyl)-5-(3-methoxyphenyl)-1,2,4-oxadiazole
Formula: C₁₀H₉ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00084966
CAS Number: 175205-62-6
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR