4-(Aminomethyl)tetrahydropyran 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 4-(Aminomethyl)tetrahydro-2H-pyran, 4-Aminomethyltetrahydropyran, Tetrahydro-2H-pyran-4-ylmethanamine, (Tetrahydro-2H-pyran-4-yl)methanamine

OR10333-5G OR10333-25G OR10333-100G
APOSOR10333-5GEA 15.8 GBP
APOSOR10333-5G APOSOR10333-25G APOSOR10333-100G
4-(Aminomethyl)tetrahydropyran 97%
4-(Aminomethyl)tetrahydropyran
Formula: C₆H₁₃NO
Boiling Pt: 75…76 °C/18mmHg
Density: 1.02 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD02179435
CAS Number: 130290-79-8
UN: 2735
ADR: 8,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR