1-(4-Methoxybenzoyl)piperazine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR0809-1G
APOSOR0809-1GEA 42 GBP
APOSOR0809-1G
1-(4-Methoxybenzoyl)piperazine
1-(4-Methoxybenzoyl)piperazine
Formula: C₁₂H₁₆N₂O₂
MW: 220.268 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02256020
CAS Number: 94747-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR