4-(Chloromethyl)-6-hydroxy-2-phenylpyrimidine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR0169-250MG
APOSOR0169-250MGEA 79.8 GBP
APOSOR0169-250MG
4-(Chloromethyl)-6-hydroxy-2-phenylpyrimidine
4-(Chloromethyl)-6-hydroxy-2-phenylpyrimidine
Formula: C₁₁H₉ClN₂O
Melting Pt: 195…198 °C
Storage Temperature: Ambient
MDL Number: MFCD00113662
CAS Number: 35252-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR