Sodium thiophosphate hydrate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

IN3322-25G IN3322-1G IN3322-5G
APOSIN3322-25GEA 184 GBP
APOSIN3322-25G APOSIN3322-1G APOSIN3322-5G
Sodium thiophosphate hydrate
Sodium thiophosphate hydrate
Formula: H₂Na₃O₄PS
Melting Pt: 56…58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041513
CAS Number: 10489-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR