Ruthenium(III) chloride hydrate 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Ruthenium(III) chloride xhydrate, Ruthenium trichloride hydrate

IN3062-5G IN3062-1G IN3062-25G
APOSIN3062-5GEA 99.9 GBP
APOSIN3062-5G APOSIN3062-1G APOSIN3062-25G
Ruthenium(III) chloride hydrate 97%
Ruthenium (III) chloride hydrate
Formula: Cl₃Ru·xH₂O
Melting Pt: 100 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149844
CAS Number: 14898-67-0
EINECS: 233-167-5
UN: 3260
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR