Ruthenium 99,95%, sponge

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

IN3058-10G
APOSIN3058-10GEA 578 GBP
APOSIN3058-10G
Ruthenium 99,95%, sponge
Ruthenium
Formula: Ru
MW: 101.07 g/mol
Boiling Pt: 4900 °C
Melting Pt: 2500 °C
Density: 12.45 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011207
CAS Number: 7440-18-8
UN: 3089
ADR: 4.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR