Benzylidenebis(tricyclohexylphosphine)dichlororuthenium 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Bis(tricyclohexylphosphine)benzylidine ruthenium(IV) dichloride

IN3049-1G
APOSIN3049-1GEA 48.3 GBP
APOSIN3049-1G
Benzylidenebis(tricyclohexylphosphine)dichlororuthenium 95%
Benzylidenebis(tricyclohexylphosphine)dichlororuthenium
Formula: C₄₃H₇₂Cl₂P₂Ru
MW: 822.9553 g/mol
Melting Pt: 153 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01090946
CAS Number: 172222-30-9
UN: 1325
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR