Potassium iodide

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

IN2918-1KG IN2918-250G
APOSIN2918-1KGEA 132 GBP
APOSIN2918-1KG APOSIN2918-250G
Potassium iodide
Potassium iodide
Formula: Kl
Boiling Pt: 1330 °C
Melting Pt: 681 °C
Density: 3.13 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR