Palladium(II) acetate trimer

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Hexakis(acetato)tripalladium(II), Tripalladium(II) Hexaacetate

IN2803-1G
APOSIN2803-1GEA 126 GBP
APOSIN2803-1G
Palladium(II) acetate trimer
Palladium(II) acetate trimer
Formula: C₁₂H₁₈O₁₂Pd₃
MW: 673.52 g/mol
Melting Pt: 205 °C
Storage Temperature: Ambient
CAS Number: 53189-26-7
EINECS: 258-424-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR