Magnesium oxide 99.99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Magnesia usta

IN2467-100G IN2467-25G
APOSIN2467-100GEA 687 GBP
APOSIN2467-100G APOSIN2467-25G
Magnesium oxide 99.99%
Magnesium oxide
Formula: MgO
MW: 40.3 g/mol
Boiling Pt: 3600 °C
Melting Pt: 2800 °C
Density: 3.58 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011109
CAS Number: 1309-48-4
Merck Index: 13,05700

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR