Lithium chloride, anhydrous 99,95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

IN2326-100G IN2326-25G IN2326-250G
APOSIN2326-100GEA 189 GBP
APOSIN2326-100G APOSIN2326-25G APOSIN2326-250G
Lithium chloride, anhydrous 99,95%
Lithium chloride
Formula: LiCl
Boiling Pt: 1325…1360 °C
Melting Pt: 605 °C
Density: 2.068 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR