Holmium trichloride hexahydrate 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

IN1975-100G IN1975-25G
APOSIN1975-100GEA 210 GBP
APOSIN1975-100G APOSIN1975-25G
Holmium trichloride hexahydrate 98%
Holmium trichloride hexahydrate
Formula: Cl₃Ho·6H₂O
Melting Pt: 165 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149739
CAS Number: 14914-84-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR