Cadmium(II) telluride 99.99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Cadmium telluride

IN1369-100G IN1369-25G
APOSIN1369-100GEA 195 GBP
APOSIN1369-100G APOSIN1369-25G
Cadmium(II) telluride 99.99%
Cadmium (II) telluride
Formula: CdTe
Melting Pt: 1041 °C
Density: 6.2 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00015998
CAS Number: 1306-25-8
EINECS: 215-149-9
UN: 2570
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR