Bismuth(III) oxide 99.999%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Bismuth Trioxide, Dibismuth trioxide

IN1255-100G IN1255-500G
APOSIN1255-100GEA 116 GBP
APOSIN1255-100G APOSIN1255-500G
Bismuth(III) oxide 99.999%
Bismuth (III) oxide
Formula: BiO₃
Boiling Pt: 1890 °C
Melting Pt: 817 °C
Density: 8.9 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003462
CAS Number: 1304-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR