Potassium tetrachloroplatinate(II) 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

IN1004-1G IN1004-5G
APOSIN1004-1GEA 52.5 GBP
APOSIN1004-1G APOSIN1004-5G
Potassium tetrachloroplatinate(II) 95%
Potassium tetrachloroplatinate (II)
Formula: Cl₄K₂Pt
Density: 3.38 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011378
CAS Number: 10025-99-7
EINECS: 233-050-9
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR