Thiamine hydrochloride ≥99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Aneurine hydrochloride, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, Thiaminium chloride, VitamineB1 hydrochloride

BIT0614-100G BIT0614-1KG BIT0614-250G
APOSBIT0614-100GEA 27.3 GBP
APOSBIT0614-100G APOSBIT0614-1KG APOSBIT0614-250G
Thiamine hydrochloride ≥99%
Thiamine hydrochloride
Formula: C₁₂H₁₈Cl₂N₄OS
Melting Pt: 247…249 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012780
CAS Number: 67-03-8
EINECS: 200-641-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR