Sorafenib (BAY 43-9006)

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BISN0134-500MG BISN0134-1G BISN0134-5G
APOSBISN0134-500MGEA 36.8 GBP
APOSBISN0134-500MG APOSBISN0134-1G APOSBISN0134-5G
Sorafenib (BAY 43-9006)
Sorafenib (BAY 43-9006)

Sorafenib is a multikinase inhibitor of Raf-1 B-Raf and VEGFR-2 with IC50 of 6 nM 22 nM and 90 nM in cell-free assays respectively.

Formula: C₂₁H₁₆ClF₃N₄O₃
Melting Pt: 197 °C
Density: 1.46 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD06411450
CAS Number: 284461-73-0
UN: 3077
ADR: 9+EHS,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR