Spermidine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine

BIS5620-25G BIS5620-5G BIS5620-50G
APOSBIS5620-25GEA 227 GBP
APOSBIS5620-25G APOSBIS5620-5G APOSBIS5620-50G
Spermidine
Spermidine

Used in the isolation of chromosomes.

Formula: C₇H₁₉N₃
MW: 145.25 g/mol
Boiling Pt: 246
Melting Pt: 24 °C
Density: 0.925 g/cm³ (25 °C)
Flash Pt: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008229
CAS Number: 124-20-9
UN: 2735
ADR: 8 II
Merck Index: 12,08893

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR