Azido-PEG7-amine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BIPG1885-1G
APOSBIPG1885-1GEA 552 GBP
APOSBIPG1885-1G
Azido-PEG7-amine
Azido-PEG7-amine
Formula: C₁₆H₃₄N₄O₇
Storage Temperature: Ambient
CAS Number: 1333154-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR