Nile red

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one

BIN0465-1G BIN0465-250MG BIN0465-5G
APOSBIN0465-1GEA 103 GBP
APOSBIN0465-1G APOSBIN0465-250MG APOSBIN0465-5G
Nile red
Nile red
Formula: C₂₀H₁₈N₂O₂
Melting Pt: 203…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00011639
CAS Number: 7385-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR