Neomycin sulphate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Neomycin, (2S,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-(((2R,3S,4R,5S)-5-(((1R,2R,3S,5R,6S)-3,5-diamino-2-(((2R,3R,4R,5S,6R)-3-amino-6-(aminomethyl)-4,5-dihydroxytetrahydro-2H-pyran-2-yl)oxy)-6-hydroxycyclohexyl)oxy)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)oxy)tetrahydro-2H-pyran-3,4-diol sulphate, Fradiomycin sulphate, Neomycin sulphate salt

BIN0135-100G BIN0135-25G
APOSBIN0135-100GEA 60.9 GBP
APOSBIN0135-100G APOSBIN0135-25G
Neomycin sulphate
Neomycin sulphate

Neomycin is an aminoglycoside antibiotic and has a bactericidal action against many gram-negative bacteria.


  • Potency: 710mg/mg
  • Neomycin B: 85%
  • Neomycin C: 10%
  • Cell culture tested

Formula: C₂₃H₄₆N₆O₁₃·3H₂SO₄
MW: 908.87 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150827
CAS Number: 1405-10-3
EINECS: 215-773-1
Merck Index: 12,06542

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR