Nalidixic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

BIN0134-100G BIN0134-25G
APOSBIN0134-100GEA 78.7 GBP
APOSBIN0134-100G APOSBIN0134-25G
Nalidixic acid
Nalidixic acid

Active against gram-negative bacteria.

Formula: C₁₂H₁₂N₂O₃
Melting Pt: 225…230 °C
Storage Temperature: Ambient
MDL Number: MFCD00006884
CAS Number: 389-08-2
EINECS: 206-864-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR