Genistein ≥98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one, 4',6'-Dihydroxy-3-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',5,7-Trihydroxyisoflavone

BIMB2006-1G BIMB2006-500MG
APOSBIMB2006-1GEA 157 GBP
APOSBIMB2006-1G APOSBIMB2006-500MG
Genistein ≥98%
Genistein

Inhibitor of tyrosine protein kinase.

Formula: C₁₅H₁₀O₅
Melting Pt: 297…298 °C
Storage Temperature: Freezer
MDL Number: MFCD00016952
CAS Number: 446-72-0
EINECS: 207-174-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR