5-Bromo-3-indolyl-β-D-galactopyranoside

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Bluo-Gal, (2S,3R,4S,5R,6R)-2-((5-Bromo-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, 5-Bromo-3-indolyl β-D-galactopyranoside

BIMB1030-100MG BIMB1030-1G BIMB1030-5G
APOSBIMB1030-100MGEA 15.8 GBP
APOSBIMB1030-100MG APOSBIMB1030-1G APOSBIMB1030-5G
5-Bromo-3-indolyl-β-D-galactopyranoside
5-Bromo-3-indolyl-beta-D-galactopyranoside

Chromogenic substrate for b-Galactosidase. Reported to produce a deeper blue water insoluble colouration than X-Gal.

Formula: C₁₄H₁₆BrNO₆
Storage Temperature: Freezer
MDL Number: MFCD00063691
CAS Number: 97753-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR