IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99% dioxane-free

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

BIMB1008-25G BIMB1008-5G BIMB1008-10G
APOSBIMB1008-25GEA 63 GBP
APOSBIMB1008-25G APOSBIMB1008-5G APOSBIMB1008-10G
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99% dioxane-free
IPTG (Isopropyl-β-D-thiogalactopyranoside)

IPTG is used in conjunction with X-Gal for detection of lac+ colonies or cells in a colorimetric assay to detect recombinants (white) from non-recombinants (blue) in cloning strategies.

Formula: C₉H₁₈O₅S
Melting Pt: 105 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063273
CAS Number: 367-93-1
EINECS: 206-703-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR