Ketotifen fumarate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 4-(1-Methylpiperidin-4-ylidene)-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-10(9H)-one fumarate

BIK4000-1G BIK4000-250MG
APOSBIK4000-1GEA 18.9 GBP
APOSBIK4000-1G APOSBIK4000-250MG
Ketotifen fumarate
Ketotifen fumarate
Formula: C₁₉H₁₉NOS
Melting Pt: 192 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00079394
CAS Number: 34580-14-8
EINECS: 252-100-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR