Hygromycin B, powder

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol

BIH4302-1G
APOSBIH4302-1GEA 174 GBP
APOSBIH4302-1G
Hygromycin B, powder
Hygromycin B

Toxic aminoglycoside produced by Streptomyces hygroscopicus. Used as a selective agent for the incorporation of the APH-4 gene in plant tissue.


  • Average Activity: 1000 units/mg
  • Cell culture tested

Formula: C₂₀H₃₇N₃O₁₃
Melting Pt: 160…180 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD06795479
CAS Number: 31282-04-9
UN: 3462
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR