L(+)-Histidine ≥99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, H-His-OH, NSC 137773, L-Histidine

BIH0710-100G BIH0710-500G
APOSBIH0710-100GEA 15.8 GBP
APOSBIH0710-100G APOSBIH0710-500G
L(+)-Histidine ≥99%
L(+)-Histidine

USP, Pyrogen free

Formula: C₆H₉N₃O₂
Melting Pt: 282 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064315
CAS Number: 71-00-1
EINECS: 200-745-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR