EPPS (HEPPS, 4-(2-hydroxyethyl)-1-piperazinepropanesulphonic acid) ≥99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

BIH0601-250G BIH0601-100G BIH0601-500G
APOSBIH0601-250GEA 126 GBP
APOSBIH0601-250G APOSBIH0601-100G APOSBIH0601-500G
EPPS (HEPPS, 4-(2-hydroxyethyl)-1-piperazinepropanesulphonic acid) ≥99%
EPPS (HEPPS, 4-(2-Hydroxyethyl)-1-piperazinepropanesulphonic acid)

Biological buffer - useful range 7.3-8.7

Formula: C₉H₂₀N₂O₄S
Melting Pt: 237…239 °C
Storage Temperature: Ambient
MDL Number: MFCD00006160
CAS Number: 16052-06-5
EINECS: 240-198-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR